Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Eumulgin™ B25

Catalog No. 7691931KG Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
76-919-31KG 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 76-919-31KG Supplier Sigma Aldrich Supplier No. 7691931KG
Only null left
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Eumulgin(R) B 25 is an oil-in-water emulsifier that is used for the production of ointments, creams, liquid emulsions, gels and other cosmetic formulations. It is supplied in the form of free-flowing, non-dusting micro-granules, and is an efficient protective colloid that can increase the stability of dispersed systems. This research grade product is intended for use in .Eumulgin is a registered trademark of Cognis IP Management GmbH

Specifications

CAS 68439-49-6
Quantity 1 kg
MDL Number MFCD03098567
Synonym Macrogol (25)-cetostearyl ether; Polyethylene glycol 1100 mono(hexadecyl/octadecyl) ether
Percent Purity ~100%active ingredients basis
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.