Learn More
Bioworld Euonymus europaeus (Spindle tree) Lectin (EEL/EEA) - FITC
Supplier:  Bioworld 217610211
Encompass_Preferred
Technical Specifications:Ligand Source:Euonymus europaeusMolecular Weight: 140 kDaCarbohydrate Specificity:Galα1,3Galβ4GlcNAc, Fucα1,2(Galα1,3)Galβ1,4GlcNAcInhibiting/Eluting Sugar:LactoseCarbohydrate for Elution: 0.2MLactoseBlood Group Specificity: O (-SA), BConjugate: FITCForm: LiquidIsoelectric (pI): 4.4Storage: -20°CApplication note: Calcium and Zinc are required for binding.
Catalog No. 50-199-127
Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please to view product availability in your area.