Learn More
Bioworld Euonymus europaeus (Spindle tree) Lectin (EEL/EEA) - Texas Red
Supplier:  Bioworld 217610231
Encompass_Preferred
Technical Specifications:Ligand Source:Euonymus europaeusMolecular Weight:140 kDaCarbohydrate Specificity:Galα1,3Galβ4GlcNAc, Fucα1,2(Galα1,3)Galβ1,4GlcNAcInhibiting/Eluting Sugar:LactoseCarbohydrate for Elution:0.2MLactoseBlood Group Specificity:O (-SA), BConjugate: Texas RedForm:LiquidIsoelectric (pI):4.4Storage: -20°CApplication note: Calcium and Zinc are required for binding.
Catalog No. 50-199-129