Learn More
Bioworld EUONYMUS EUROPAEUS LECTIN,2MG
Supplier:  Bioworld 215100541
Encompass_Preferred
Euonymus europaeus Lectin (EEL/EEA) - Biotinylated.Specifications:Source Euonymus europaeus (Spindle tree)Activity: 50 mg/ml will agglutinate human A2 erythrocytes.Carbohydrate Specificity: Galα3GalInhibitory Carbohydrate: LactoseDivalent Ions Required: Ca++, Zn++Conjugation: Biotin
Catalog No. NC2119082