Learn More
Bioworld EUONYMUS EUROPAEUSSP TREE
Supplier:  Bioworld 215100041
Encompass_Preferred
EUONYMUS EUROPAEUS (SP TREE) - LECTIN: Euonymus europaeus (spindle tree) Lectin isolated from Euonymus europaeus (spindle tree) seeds. Specifications: Primary sugar specificity: Galactose Preferential binding activity: galactosyl ...
Catalog No. 50-198-735
Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please to view product availability in your area.