Learn More
Cayman Chemical Eupalinolide A, Purity: ≥98%, CAS: 877822-40-7, 10 mg
SDP
Catalog No. 502947705
Encompass_Preferred
A sesquiterpene lactone with cellular chaperone-inducing activity; activates Hsf1 and induces the expression and increases the protein levels of Hsp70 in B16 cells at 5, 7.5, and 10 µg/ml
Catalog No. 50-294-7705 Supplier Cayman Chemical Supplier No. 3380810 MG
A sesquiterpene lactone with cellular chaperone-inducing activity; activates Hsf1 and induces the expression and increases the protein levels of Hsp70 in B16 cells at 5, 7.5, and 10 µg/ml