Learn More
Medchemexpress LLC HY-N0753 5mg Medchemexpress, Eupalinolide B CAS:877822-41-8 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYN07535MG
Encompass_Preferred
Medchemexpress, HY-N0753 5mg Eupalinolide B CAS:877822-41-8 Eupalinolide B is a germacrane sesquiterpene isolated from Eupatorium lindleyanum. Eupalinolide B demonstrates potent cytotoxicity against A-549, BGC-823 and HL-60 tumour cell lines. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2817