Learn More
TARGETMOL CHEMICALS INC Eupatilin 10MG
SDP
Catalog No. 502878008
Encompass_Preferred
Also available in 1 mL, 1 mg, 5 mg, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. Eupatilin (NSC-122413), a flavone derived from Artemisia princepsPampanini, is a PPARalpha agonist, with pharmacological activity. Purity 98.53%
Catalog No. 50-287-8008 Supplier TARGETMOL CHEMICALS INC Supplier No. T336210MG
Also available in 1 mL, 1 mg, 5 mg, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. Eupatilin (NSC-122413), a flavone derived from Artemisia princepsPampanini, is a PPARalpha agonist, with pharmacological activity. Purity 98.53%