Learn More
Medchemexpress LLC HY-N0783 10mg Medchemexpress, Eupatilin CAS:22368-21-4 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYN078310MG
Encompass_Preferred
Medchemexpress, HY-N0783 10mg Eupatilin CAS:22368-21-4 Eupatilin, a lipophilic flavonoid isolated from Artemisia species, is a PPARα agonist, and possesses anti-apoptotic, anti-oxidative and anti-inflammatory activities. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-3715