Learn More
Medchemexpress LLC Eupatilin | 22368-21-4 | 344.32 | 500 MG
SDP
Catalog No. 5000417181
Encompass_Preferred
Eupatilin is a lipophilic flavonoid isolated from *Artemisia argyi Lévl. et Van.* It acts as a PPARα agonist and exhibits anti-apoptotic, anti-oxidative, and anti-inflammatory activities.
  • Acts as a PPARα agonist
  • Demonstrates anti-apoptotic activities
  • Exhibits anti-oxidative properties
  • Shows anti-inflammatory activities
Catalog No. 50-004-17181 Supplier Medchemexpress LLC Supplier No. HYN0783500MG
Eupatilin is a lipophilic flavonoid isolated from *Artemisia argyi Lévl. et Van.* It acts as a PPARα agonist and exhibits anti-apoptotic, anti-oxidative, and anti-inflammatory activities.
  • Acts as a PPARα agonist
  • Demonstrates anti-apoptotic activities
  • Exhibits anti-oxidative properties
  • Shows anti-inflammatory activities