Learn More
Selleck Chemical LLC Eupatilin S3846-5mg
SDP
Catalog No. 503109808
Encompass_Preferred
Eupatilin (NSC 122413) a major flavonoid from Artemisia plants possesses various beneficial biological effects including anti-inflammation anti-tumor anti-cancer anti-allergy and anti-oxidation activity Eupatilin a lipophilic flavonoid isolated from Artemisia species is a PPAR agonist and possesses anti-apoptotic anti-oxidative and anti-inflammatory activities
Catalog No. 50-310-9808 Supplier Selleck Chemical LLC Supplier No. S38465MG
Eupatilin (NSC 122413) a major flavonoid from Artemisia plants possesses various beneficial biological effects including anti-inflammation anti-tumor anti-cancer anti-allergy and anti-oxidation activity Eupatilin a lipophilic flavonoid isolated from Artemisia species is a PPAR agonist and possesses anti-apoptotic anti-oxidative and anti-inflammatory activities