Learn More
Cayman Chemical EupenIfeldIn 100ug
SDP
Catalog No. 502933560
Encompass_Preferred
A cytotoxic fungal metabolite (IC50s = 0.005 and 0.002 μg/ml for HCT116 and HCTVM46 cells, respectively); increases median survival time in a mouse model of P388 leukemia from 0.3-2 mg/kg per day
Catalog No. 50-293-3560 Supplier Cayman Chemical Supplier No. 25114100 UG
A cytotoxic fungal metabolite (IC50s = 0.005 and 0.002 μg/ml for HCT116 and HCTVM46 cells, respectively); increases median survival time in a mouse model of P388 leukemia from 0.3-2 mg/kg per day