Learn More
Medchemexpress LLC HY-N0313 5mg Medchemexpress, Euphol CAS:514-47-6 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYN03135MG
Encompass_Preferred
Medchemexpress, HY-N0313 5mg Euphol CAS:514-47-6 Euphol is a tetracyclic triterpene alcohol isolated from the sap of Euphorbia tirucalli with anti-mutagenic, anti-inflammatory and immunomodulatory effects, orally active. Euphol inhibits the monoacylglycerol lipase (MGL) activity via a reversible mechanism (IC50=315 nM). MGL inhibition in the periphery modulates the endocannabinoid system to block the development of inflammatory pain. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-197-0251