Learn More
Cayman Chemical Euphorbiasteroid, Purity: ≥98%, CAS: 28649-59-4, 100 mg
SDP
Catalog No. 502941684
Encompass_Preferred
A tricyclic diterpene; inhibits early-stage adipogenesis of 3T3-L1 cells at 25 and 50 µM; inhibits proliferation of HL-60 cells in a concentration-dependent manner, as well as induces apoptosis and increases the expression of Fas and Fas ligand (FasL) and the activity of caspase-3 and caspase-8 in HL-60 cells; increases P-glycoprotein activity, reverses multi-drug resistance, and restores cytotoxicity of vinblastine, paclitaxel, and doxorubicin to MES/SA/Dx5 sarcoma cells
Catalog No. 50-294-1684 Supplier Cayman Chemical Supplier No. 27822100 MG
A tricyclic diterpene; inhibits early-stage adipogenesis of 3T3-L1 cells at 25 and 50 µM; inhibits proliferation of HL-60 cells in a concentration-dependent manner, as well as induces apoptosis and increases the expression of Fas and Fas ligand (FasL) and the activity of caspase-3 and caspase-8 in HL-60 cells; increases P-glycoprotein activity, reverses multi-drug resistance, and restores cytotoxicity of vinblastine, paclitaxel, and doxorubicin to MES/SA/Dx5 sarcoma cells