Learn More
Ace Glass, Inc. European flask, 500mL, 3 neck, 24/40 center, (2) 24/40 sides
SDP
Catalog No. 50973933
Encompass_Preferred
European style 3-neck 500mL flask with tapered walls and a shallow bottom for more efficient magnetic or mechanical half-moon shaped paddle stirring. Flask has a 24/40 center neck, and (2) 24/40 side necks. Easy to clean.
Catalog No. 50-973-933 Supplier Ace Glass, Inc. Supplier No. 696116
European style 3-neck 500mL flask with tapered walls and a shallow bottom for more efficient magnetic or mechanical half-moon shaped paddle stirring. Flask has a 24/40 center neck, and (2) 24/40 side necks. Easy to clean.