Learn More
Ace Glass, Inc. European flask, jacketed, 250mL, 5 neck, 29/42 center, (2)24/40, (2) #7 Ace-Thred
SDP
Catalog No. 50973926
Encompass
Catalog No. 50-973-926 Supplier Ace Glass, Inc. Supplier No. 695951