Learn More
Ace Glass, Inc. European flask, 500mL, 5 neck, 24/40 center, (2) 24/40, (2)#7 Ace-Thred
SDP
Catalog No. 50973939
Encompass_Preferred
European style 5-neck 500mL flask with tapered walls and a shallow bottom for more efficient magnetic or mechanical half-moon shaped paddle stirring. Flask has a 24/40 center neck, (2) 24/40 side necks, and (2) #7 Ace-Thred necks. Includes nylon bushings and FETFE o-rings for the Ace-Threds to be used with thermometers, etc. Easy to clean.
Catalog No. 50-973-939 Supplier Ace Glass, Inc. Supplier No. 696175
European style 5-neck 500mL flask with tapered walls and a shallow bottom for more efficient magnetic or mechanical half-moon shaped paddle stirring. Flask has a 24/40 center neck, (2) 24/40 side necks, and (2) #7 Ace-Thred necks. Includes nylon bushings and FETFE o-rings for the Ace-Threds to be used with thermometers, etc. Easy to clean.