Learn More
GSC Go Science Crazy Euro Style Polypropylene Beakers
SCI_ED
Catalog No. S13803 Shop All GSC Products
Click to view available options
Capacity (Metric):
1000 mL
2000 mL
250 mL
50 mL
500 mL
Assorted
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric)
S13803 Assorted
S13797 50 mL
S13799 250 mL
S13800 500 mL
S13802 2000 mL
S13801 1000 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Intended for ages 13 and up.
Catalog No. S13803 Supplier GSC Go Science Crazy Supplier No. BKPPASR
Intended for ages 13 and up.
Incorporate raised outer graduations for ease of reading. GSC International Euro Style Polypropylene Beakers are tough, chemical resistant, and autoclavable. They are great additions to laboratory or classroom settings.
  • Molded from tough translucent polypropylene
  • Incorporate raised outer graduations for ease of reading
  • Tapered spout for easy pouring
  • Chemical resistant
  • Autoclavable
Specifications
Color Translucent
Capacity (Metric) Assorted
Material Polypropylene
Includes Assorted - One each of volumes: 50mL, 100mL, 250mL, 500mL, 1000mL
Autoclavable Autoclavable
Graduated Yes
Scale Raised Scale
Style Euro
WARNING: Cancer - www.P65Warnings.ca.gov