Learn More
Description
- Ideal for life and materials sciences, various industrial applications and for stereo microscopy
- Light source with two self-sustaining gooseneck-type guides, equipped with 2x 3W white 6500 K power LED.
- Each self-sustaining gooseneck-type LED guide is equipped with a 3-lens focusing head and has a length of 56 cm
- The light source version does not require bulb changing like halogen light sources and has a long-ife span and low energy consumption
- The light source produces white light and there is no color temperature shifting when dimming
- Evenly illuminates specimen without heating
- 2-year warranty
Specifications
Specifications
| Type | Euromex Dual LED Illuminator Cold Light Source |
| Autoclavable | Not Autoclavable |
| Sterility | Non-sterile |
| Voltage | 110/220 V |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.