Description
- All Globe Euromex cameras are supplied with photo port c-mount adapters, which means they can be connected to any Globe Euromex trinocular microscope*.
- * Except for the Delphi-X Inverso series. These microscopes need an additional adapter.
Photo Port Adapters
- Suitable for Globe Euromex microscopes with a photo port such as Oxion Inverso, NexiusZoom, StereoBlue and Delphi-X Observer
Specifications
Specifications
| Type | Photo Port Adapter |
| Autoclavable | Not Autoclavable |
| For Use With (Equipment) | Model EOX-2053-PLPH and cameras EDC-5000-PRO / EDC-10000-PRO / EDC-18000-PRO / EVC-3042 |
| Sterility | Non-sterile |
| Voltage | 110/220 V |