Learn More
Globe Scientific Euromex T2 Ring for SLR Digital Camera
Catalog No. 23556101
Change view
Click to view available options
For Use With (Equipment):
Canon EOS SLR Digital Camera
Nikon D SLR Digital Camera
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
23-556-101 Nikon D SLR Digital Camera
23-556-102 Canon EOS SLR Digital Camera
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 23-556-101 Supplier Globe Scientific Supplier No. EAE5025
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy
Digital single lens reflex cameras with an APS-C sensor can be mounted on a standard 23.2 mm trinocular tube. Choose the universal EAE-5130 adapter with built-in 2x objective, together with a brand-specific T-2 adapter.
  • All Globe Euromex cameras are supplied with photo port c-mount adapters, which means they can be connected to any Globe | Euromex trinocular microscope*.
    • * Except for the Delphi-X Inverso series. These microscopes need an additional adapter.
Digital SLR Adapters
  • Digital single lens reflex cameras with an APS-C sensor can be mounted on a standard 23.2mm trinocular tube.
  • Choose the universal EAE-5130 adapter with built-in 2x objective, together with a brand-specific T-2 adapter.
Specifications
Type Euromex T2 Ring for Nikon D SLR Digital Camera
Autoclavable Not Autoclavable
For Use With (Equipment) Nikon D SLR Digital Camera
Sterility Non-sterile
Voltage 110/220 V