Description
- All Globe Euromex cameras are supplied with photo port c-mount adapters, which means they can be connected to any Globe | Euromex trinocular microscope*.
- * Except for the Delphi-X Inverso series. These microscopes need an additional adapter.
Digital SLR Adapters
- Digital single lens reflex cameras with an APS-C sensor can be mounted on a standard 23.2mm trinocular tube.
- Choose the universal EAE-5130 adapter with built-in 2x objective, together with a brand-specific T-2 adapter.
Specifications
Specifications
| Type | Euormex T2 Ring for Canon EOS SLR Digital Camera |
| Autoclavable | Not Autoclavable |
| For Use With (Equipment) | Canon EOS SLR Digital Camera |
| Sterility | Non-sterile |
| Voltage | 110/220 V |