Description
- Ultra HD 2160p, 8.3 MP, 1/8 in. color 4K Sony sensor
- 3840 x 2160 pixels, 30 fps, H264 codec
- Real time images directly on TV, monitor or beamer
- Built-in mouse-driven software
- 4K HDMI 2.0 standard
- Interface: HDMI, USB-2, Ethernet, Wi-Fi
- Works with ImageFocus Alpha software
- ADC 16 bits, color depth 36 bits
- Stand alone or used via PC
- C-mount interface
- 2061p/1080p automatic switching
- SD memory card included
Specifications
Specifications
| Type | Camera |
| Autoclavable | Not Autoclavable |
| Camera Type | UHD-4K Camera |
| Sterility | Non-sterile |
| Voltage | 110/220 V |