Learn More
Sigma Aldrich Fine Chemicals Biosciences Europine hydrochloride phy10MG
Catalog No. 502908452
Encompass
This substance is a primary reference substance with assigned absolute purity (considering chromatographic purity water residual solvents inorganic impurities). The exact value can be found on the certificate. Produced by PhytoLab GmbH Co. KG
Catalog No. 50-290-8452 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. PHL8323710MG
This substance is a primary reference substance with assigned absolute purity (considering chromatographic purity water residual solvents inorganic impurities). The exact value can be found on the certificate. Produced by PhytoLab GmbH Co. KG