Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Europium(III) acetate hydrate, REacton™, 99.9% (REO)

Catalog No. AA4287806
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4287806 5 g
AA4287814 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA4287806 Supplier Thermo Scientific Chemicals Supplier No. 04287806
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 62667-64-5 | C6H9EuO6 | 329.10 g/mol

Europium(III) acetate hydrate is used as fluorescent powder and as a chemical reagent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 62667-64-5
Molecular Formula C6H9EuO6
Molecular Weight (g/mol) 329.10
MDL Number MFCD00150115
InChI Key LNYNHRRKSYMFHF-UHFFFAOYSA-K
Synonym europium iii acetate hydrate 1:3:x, europium acetate hydrate, europium acetate,hydrate, europium iii acetatehydrate, c6h9euo6.h2o, europium iii acetate hydrate, reacton, europium iii acetate hydrate, reacton?, bis acetyloxy europio acetate hydrate, europium iii acetate hydrate trace metals basis
PubChem CID 16212055
SMILES [Eu+3].CC([O-])=O.CC([O-])=O.CC([O-])=O

Specifications

Color White
Physical Form Powder
Quantity 5 g
Assay Percent Range 99.9% (REO)
Solubility Information Soluble in water.
IUPAC Name europium(3+) triacetate
Formula Weight 329.10 (Anhydrous)
Sensitivity Hygroscopic
Appearance White crystalline powder
Chemical Name or Material Europium(III) acetate hydrate
EINECSNumber 214-670-9
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.