Learn More
Strem, An Ascensus Company CAS# 13759-92-7. 25g. Europium(III) chloride hexahydrate (99.9%-Eu) (REO). MFCD00149701
SDP
Catalog No. 5090116028
Encompass
CAS# 13759-92-7. 25g. Europium(III) chloride hexahydrate (99.9%-Eu) (REO). MFCD00149701. Molecular Weight: 258.32 (366.41). Molecular Formula: EuCl3∙6H2O. Color/form: white to off-white xtl. Strem# 93-6306. http://www.strem.com/catalog/v/93-6306/
Catalog No. 50-901-16028 Supplier Strem, An Ascensus Company Supplier No. 93630625G
CAS# 13759-92-7. 25g. Europium(III) chloride hexahydrate (99.9%-Eu) (REO). MFCD00149701. Molecular Weight: 258.32 (366.41). Molecular Formula: EuCl3∙6H2O. Color/form: white to off-white xtl. Strem# 93-6306. http://www.strem.com/catalog/v/93-6306/