Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Europium(III) chloride hydrate, REacton™, 99.9% (REO)

Catalog No. AA1129806
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA1129806 5 g
AA1129814 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA1129806 Supplier Thermo Scientific Chemicals Supplier No. 01129806
Add to Cart

missing translation for 'validMainframeMsg'

EuCl3·xH2O

Europium(III) chloride hydrate is used as a catalyst in organic synthesis. It is involved in the preparation of amino acid based europium(III) complex in order to prepare photoactive thin films, which finds application in organic light emitting diodes. It is also used as a precursor for the preparation of other europium compounds and its salts.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

Molecular Formula EuCl3·xH2O
MDL Number MFCD00010994

Specifications

Color Yellow
Physical Form Crystalline Aggregates
Quantity 5 g
Assay Percent Range 99.9% (REO)
Solubility Information Soluble in water.
Formula Weight 258.32 (anhydrous)
Odor Odorless
Sensitivity Hygroscopic
Chemical Name or Material Europium(III) chloride hydrate
EINECSNumber 233-040-4
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.