Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Europium(III) fluoride, anhydrous, REacton™, 99.98% (REO)

Catalog No. AA1130204
Change view
Click to view available options
Quantity:
2 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA1130204 2 g
AA1130206 5 g
AA1130214 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA1130204 Supplier Thermo Scientific Chemicals Supplier No. 01130204
Add to Cart

CAS: 13765-25-8 | EuF3 | 208.96 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13765-25-8
Molecular Formula EuF3
Molecular Weight (g/mol) 208.96
MDL Number MFCD00010998
InChI Key HPNURIVGONRLQI-UHFFFAOYSA-K
Synonym europium fluoride, europium iii fluoride, acmc-20akvp, europium 3+ ion trifluoride, europium iii fluoride, anhydrous trace rare earth metals basis 1g
IUPAC Name europium(3+) trifluoride
SMILES [F-].[F-].[F-].[Eu+3]

Specifications

Boiling Point >2280°C
Melting Point 1390°C
Physical Form Powder
Quantity 2 g
Formula Weight 208.96
Percent Purity 99.98%
Grade Reacton
Sensitivity Hygroscopic
Assay (REO)
Chemical Name or Material Europium(III) fluoride, Anhydrous
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 237-368-9
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.