Learn More
Strem, An Ascensus Company CAS# 10031-53-5. 5g. Europium(III) nitrate hexahydrate (99.9%-Eu) (REO). MFCD00149700
SDP
Catalog No. 5090116032
Encompass
CAS# 10031-53-5. 5g. Europium(III) nitrate hexahydrate (99.9%-Eu) (REO). MFCD00149700. Molecular Weight: 337.98 (446.07). Molecular Formula: Eu(NO3)3∙6H2O. Color/form: white xtl. Strem# 93-6310. http://www.strem.com/catalog/v/93-6310/
Catalog No. 50-901-16032 Supplier Strem, An Ascensus Company Supplier No. 9363105G
CAS# 10031-53-5. 5g. Europium(III) nitrate hexahydrate (99.9%-Eu) (REO). MFCD00149700. Molecular Weight: 337.98 (446.07). Molecular Formula: Eu(NO3)3∙6H2O. Color/form: white xtl. Strem# 93-6310. http://www.strem.com/catalog/v/93-6310/