Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Europium(III) nitrate hydrate, REacton™, 99.99% (REO)

Catalog No. AA1129614
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA1129614 25 g
AA1129606 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA1129614 Supplier Thermo Scientific Chemicals Supplier No. 01129614
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 100587-95-9 | EuN3O9 | 337.98 g/mol

Europium(III) nitrate hydrate is widely used as dopant and used in the preparation of europium-doped hafnium and europium doped gadolinium oxides.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 100587-95-9
Molecular Formula EuN3O9
Molecular Weight (g/mol) 337.98
MDL Number MFCD00149700
InChI Key GAGGCOKRLXYWIV-UHFFFAOYSA-N
Synonym europium iii nitrate hydrate, acmc-1c6y6, eu.3no3.h2o, europium trinitrate monohydrate, europium 3+ hydrate trinitrate, europium 3+ ion hydrate trinitrate, nitric acid,europium 3+ salt, hydrate 9ci
PubChem CID 57346089
SMILES [Eu+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O

Specifications

Melting Point 85°C
Color Colorless to Pink
Physical Form Crystalline Aggregates
Quantity 25 g
UN Number UN1477
Merck Index 14,3904
Solubility Information Highly soluble in water.
IUPAC Name europium(3+) trinitrate
Formula Weight 337.97 (Anhydrous)
Percent Purity 99.99%
Grade Reacton
Assay (REO)
Chemical Name or Material Europium(III) nitrate hydrate
Show More Show Less
Hazard Category H272-H315-H319-H335
Hazard Statement GHS H Statement
H272-H315-H319-H335
May intensify fire
oxidizer.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P220-P221-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c
DOTInformation Transport Hazard Class: 5.1; Packing Group: III; Proper Shipping Name: NITRATES, INORGANIC, N.O.S.
EINECSNumber 233-380-3
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.