Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Europium(III) nitrate pentahydrate

Catalog No. 2079181G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
10 g
50 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
20-791-81G 1 g
20-791-810G 10 g
20-791-850G 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 20-791-81G Supplier Sigma Aldrich Supplier No. 2079181G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Europium(III) nitrate pentahydrate is primarily used as a dopant. It has been used to prepare Gd2O3-doped Eu (5 mol%) films. Europium-doped hafnium oxide (Eu3+:HfO2) may be prepared by a sol-gel process. Also, synthesis of europium doped zirconium carbonate nanoparticles may also be undertaken.

Specifications

CAS 63026-01-7
Molecular Formula EuN3O9 -+ 5H2O
Linear Formula Eu(NO3)3 -+ 5H2O
Quantity 1 g
MDL Number MFCD00149698
Synonym Europium trinitrate hydrate
Molecular Weight (g/mol) 428.06
Percent Purity 99.9% trace metals basis
Biological Activity Respiratory system
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.