Alfa Aesar™ Europium(III) sulfate octahydrate, 99.9% (REO)

Manufacturer: Alfa Aesar™ 2910609

Quantity 10g

 View more versions of this product

Catalog No. AA2910609

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 10031-55-7
    Formula Weight 736.23 (592.10 Anhydrous)
    MDL Number MFCD00149704
    Merck Index 14,3904
    Chemical Name or Material Europium(III) sulfate octahydrate

    Specifications

    Formula Weight 736.23 (592.10 Anhydrous)
    Melting Point 375°C (in -8H2O)
    Health Hazard 3 GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Odor Odorless
    TSCA TSCA
    Sensitivity Hygroscopic
    MDL Number MFCD00149704
    CAS 10031-55-7
    Merck Index 14,3904
    Quantity 10g
    Physical Form Crystalline
    Health Hazard 1 Warning
    Health Hazard 2 GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Chemical Name or Material Europium(III) sulfate octahydrate
    Assay 99.9%, (REO)

    GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit