Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Europium(III) thenoyltrifluoroacetonate, trihydrate, 95%

Catalog No. AC423190050
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC423190050 5 g Glass bottle
AC423190010 1 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC423190050 Supplier Thermo Scientific Chemicals Supplier No. 423190050
Add to Cart

CAS: 21392-96-1 | C24H12EuF9O6S3·3H2O | 869.54 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 21392-96-1
Molecular Formula C24H12EuF9O6S3·3H2O
Molecular Weight (g/mol) 869.54
MDL Number MFCD00150912
InChI Key LSMLVRQTTAIDBZ-BSWAEIBTSA-N
Synonym tris 4,4,4-trifluoro-1-2-thienyl-1,3-butanediono europium iii hydrate
PubChem CID 91873327
IUPAC Name erbium;(Z)-1,1,1-trifluoro-4-hydroxy-4-thiophen-2-ylbut-3-en-2-one;hydrate
SMILES C1=CSC(=C1)C(=CC(=O)C(F)(F)F)O.C1=CSC(=C1)C(=CC(=O)C(F)(F)F)O.C1=CSC(=C1)C(=CC(=O)C(F)(F)F)O.O.[Er]

Specifications

Melting Point 142.0°C to 147.0°C
Color Yellow
Elemental Analysis 94% min.
Infrared Spectrum Authentic
Assay Percent Range 95%
Packaging Glass bottle
Quantity 5 g
Solubility Information Solubility in water: insoluble
Formula Weight 869.54
Percent Purity 95%
Physical Form Crystalline Powder or Crystals
Chemical Name or Material Europium(III) thenoyltrifluoroacetonate, trihydrate, 95%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.