Learn More
Cayman Chemical EuropIum W1024 ITC 100ug
SDP
Supplier:  Cayman Chemical 400536100 UG
Encompass_Preferred
A lanthanide-based fluorophore for TR-FRET applications; ex/em = 340/613 nm; reacts with primary amino groups at an alkaline pH; has commonly been conjugated to various molecules and used as a donor fluorophore in TR-FRET
Catalog No. 50-294-2404