Learn More
Sigma Aldrich Fine Chemicals Biosciences EUROSTAR 200 control P4 ov1EA
Catalog No. 502908795
Encompass
Extremely powerful laboratory stirrer for highly viscous applications for quantities up to 100 L (H2O). It is designed with a removable wireless controller and a digital TFT display. It automatically adjusts the speed through microprocessor controlled technology within the speed range of 0/4 - 530 rpm (two speed ranges). The stirrer comes equipped with a RS 232 and a USB interface to control and document all parameters. An integrated torque trend display is provided for the measurement of viscosity changes. Safety circuits installed ensures automatic cut-off in an anti-stall or overload conditions. Continuous comparison of shaft speed to desired speed is maintained and variations are adjusted automatically. This guarantees a constant speed even with changes in viscosities of the sample..nbspMultilingual TFT display .nbspProgrammable functions .nbspIntegrated temperature measurement .nbspInterval operation .nbspTimer function .nbspAdjustable safety circuit .nbspLocked function .nbspInfi
Catalog No. 50-290-8795 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. Z7678591EA
Add to Cart
Add to Cart
Request a Quote
Extremely powerful laboratory stirrer for highly viscous applications for quantities up to 100 L (H2O). It is designed with a removable wireless controller and a digital TFT display. It automatically adjusts the speed through microprocessor controlled technology within the speed range of 0/4 - 530 rpm (two speed ranges). The stirrer comes equipped with a RS 232 and a USB interface to control and document all parameters. An integrated torque trend display is provided for the measurement of viscosity changes. Safety circuits installed ensures automatic cut-off in an anti-stall or overload conditions. Continuous comparison of shaft speed to desired speed is maintained and variations are adjusted automatically. This guarantees a constant speed even with changes in viscosities of the sample..nbspMultilingual TFT display .nbspProgrammable functions .nbspIntegrated temperature measurement .nbspInterval operation .nbspTimer function .nbspAdjustable safety circuit .nbspLocked function .nbspInfi