Learn More
Siemens Healthineers Eurotrol Epoc™ Calibration Verification Fluid
Catalog No. 22347445 Shop All Siemens Healthineers Products
Change view
Click to view available options
Level:
1 to 5
Catalog No. Level
22-347-445 1 to 5
1 options
This item is not returnable. View return policy
Catalog No. 22-347-445 Supplier Siemens Healthineers Supplier No. 10736514
Add to Cart
Add to Cart
This item is not returnable. View return policy
Siemens Healthineers Eurotrol Epoc™ Calibration Verification Fluid is an assayed blood gas, electrolyte and metabolite reference material, to verify precision, accuracy, and linearity of the epoc Blood Analysis System
  • Developed to verify calibration, operating temperature and other performance characteristics
  • pH, gases, electrolytes and metabolites in one ampule
  • Five (5) levels covering the physiologically relevant range
  • Use only one epoc cartridge for each level
  • Optimized for epoc with physiological target values
  • Now includes BUN and TCO2
TRUSTED_SUSTAINABILITY
Specifications
Level 1 to 5
Quantity 4 x 2.5 mL Samples