Learn More
Siemens Healthineers Eurotrol Epoc™ HCT Control
Catalog No. 22347435 Shop All Siemens Healthineers Products
Change view
Click to view available options
Level:
A
B
C
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Level
22-347-435 A
22-347-436 C
22-347-437 B
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 22-347-435 Supplier Siemens Healthineers Supplier No. 10736359
Add to Cart
Add to Cart
Siemens Healthineers Eurotrol Epoc™ HCT Control is an assayed quality control to verify precision and accuracy of the Siemens epoc Blood Analysis System measuring hematocrit
  • Three (3) levels covering the physiologically relevant range
  • Optimized for epoc with instrument specific target ranges
TRUSTED_SUSTAINABILITY
Specifications
Level A
Quantity 10 x 2.5 mL
WARNING: Cancer - www.P65Warnings.ca.gov