Learn More
Siemens Healthineers Eurotrol Epoc™ HCT Linearity Verification Mater
Catalog No. 22347434 Shop All Siemens Healthineers Products
Change view
Click to view available options
Level:
1 to 5
Catalog No. Level
22-347-434 1 to 5
1 options
Catalog No. 22-347-434 Supplier Siemens Healthineers Supplier No. 10736358
Add to Cart
Add to Cart
Siemens Healthineers Eurotrol Epoc™ HCT Linearity Verification Mater is an assayed hematocrit reference material to verify precision, accuracy and linearity of the Siemens epoc Blood Analysis System measuring Hematocrit
  • Five (5) levels covering the physiologically relevant range
  • Optimized for epoc with physiological target values
TRUSTED_SUSTAINABILITY
Specifications
Level 1 to 5
Quantity 4 x 2.5 mL Samples
WARNING: Cancer - www.P65Warnings.ca.gov