Learn More
Siemens Healthineers Eurotrol GAS-ISE Metabolites
Catalog No. 22347444 Shop All Siemens Healthineers Products
Change view
Click to view available options
Level:
1
2
3
6
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Level
22-347-444 6
22-347-441 1
22-347-442 2
22-347-443 3
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. 22-347-444 Supplier Siemens Healthineers Supplier No. 10736513
Add to Cart
Add to Cart
This item is not returnable. View return policy
Siemens Healthineers Eurotrol GAS-ISE Metabolites is an assayed blood gas, electrolyte and metabolite reference material to verify the precision and accuracy of the epoc Blood Analysis System
  • pH, gases, electrolytes and metabolites in one ampule
  • Three (3) levels covering the physiologically relevant range
  • Use only one epoc cartridge for each control check
  • Optimized for epoc with physiological target values
  • Developed to verify calibration, operating temperature and other performance characteristics
  • Now includes BUN and TCO2
TRUSTED_SUSTAINABILITY
Specifications
Level 6
Quantity 10 x 2.5 mL Vials