Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Innovating Science™ Eutectic Alloys Chemistry Demo Kit
GREENER_CHOICESCI_ED

Catalog No. S37749 Shop All Aldon Products
Change view
Click to view available options
Class Size:
1 Class
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Class Size
S37749 1 Class
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S37749 Supplier Innovating Science™ Supplier No. IS7014
Only null left
Add to Cart
Add to Cart

Includes

1 x 100 mL Glycerin (Glycerol), 1 x 50 g Bismuth Metal Lumps, 1 x 15 g Tin Metal Mossy

Form a metal alloy as bismuth and tin are heated to the eutectic point of the two metals which is lower than the melting point of either metal.

  • Form a metal alloy by heating bismuth and tin

Specifications

Color White
Grade 7 to 14
Age 13 to 20
For Use With (Application) Form a metal alloy by heating bismuth and tin
Includes 1 x 100 mL Glycerin (Glycerol), 1 x 50 g Bismuth Metal Lumps, 1 x 15 g Tin Metal Mossy
Quantity 1
Dimensions (L x W x H) 4.25 x 4.625 x 5.5 in.
Class Size 1 Class
Product Type Eutectic Alloys Chemistry Demo Kit

DOT: Non regulated

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.