Learn More
Euthanex Rectangle Euthanasia Lid (18 x 15) (457 x 381 mm) for Home Cage with QD Connector.
Catalog No. NC1880327
Encompass
Rectangle Euthanasia Lid for Home Cage with quick-disconnect lid stem for CO2 connection hose (EP-1130). Lid size is 18" x 15". Comes with gasket installed on bottom for a secure fit. Two holes are punched into top to allow air to be pushed out and replaced with CO2.
Catalog No. NC1880327 Supplier Euthanex Supplier No. E20045
Rectangle Euthanasia Lid for Home Cage with quick-disconnect lid stem for CO2 connection hose (EP-1130). Lid size is 18" x 15". Comes with gasket installed on bottom for a secure fit. Two holes are punched into top to allow air to be pushed out and replaced with CO2.