Learn More
Braintree Scientific Euthanex Lid,Opti Mouse Lid
SDP
Catalog No. 501954710
Encompass
Euthanex Lid - Euthanex Lid system has become an industry standard. Simply by removing the water bottle from the wire cage cover and placing Euthanex Lid over the host cage, user is ready to perform euthanasia. Eliminates the need to transfer animals. Lids are available in five sizes to accommodate virtually all-plastic cage sizes designed for small lab animals. Heavy-duty stainless steel lids have a stem for easy connections to the regulator hose. A foam lid gasket assures a good seal. Multiple lids may be used to treat several cages at once. **Sold for research purposes only and not for human use**
Catalog No. 50-195-4710 Supplier Braintree Scientific Supplier No. E20039
 Restricted Item
Add to Cart
Add to Cart
Euthanex Lid - Euthanex Lid system has become an industry standard. Simply by removing the water bottle from the wire cage cover and placing Euthanex Lid over the host cage, user is ready to perform euthanasia. Eliminates the need to transfer animals. Lids are available in five sizes to accommodate virtually all-plastic cage sizes designed for small lab animals. Heavy-duty stainless steel lids have a stem for easy connections to the regulator hose. A foam lid gasket assures a good seal. Multiple lids may be used to treat several cages at once. **Sold for research purposes only and not for human use**