Learn More
Euthanex EUTHANEX LID SMALL 16LX10W
Catalog No. NC9499175
Encompass
E-20028 Small Lid, 16" L x 10" WThe E-20028 Small Lid is constructed of heavy-duty, stainless steel and includes an easy, quick disconnect lid stem for gas input as well as a foam gasket for a secure fit. The foam gasket seal also allows the E-20028 to be used with smaller cage sizes.
Catalog No. NC9499175 Supplier Euthanex Supplier No. E20028
E-20028 Small Lid, 16" L x 10" WThe E-20028 Small Lid is constructed of heavy-duty, stainless steel and includes an easy, quick disconnect lid stem for gas input as well as a foam gasket for a secure fit. The foam gasket seal also allows the E-20028 to be used with smaller cage sizes.