Learn More
World Precision Instrument Euthanex Lids-EZ-20027
SDP
Catalog No. 502538837
Encompass
The Euthanex Lid system has become an industry standard. Simply by removing the water bottle from the wire bar cage cover and placing the Euthanex Lid over the host cage, you are ready to perform euthanasia. You no longer need to transfer animals. The lids are available in five sizes to accommodate virtually all plastic cage sizes designed for small lab animals. These heavy duty stainless steel lids have a stem fitting for connection to the quick-disconnect fitting on the regulator hose from the regulator. A foam lid gasket ensures a good seal on the cage. Multiple lids may be used to treat several cages at once.
Catalog No. 50-253-8837 Supplier World Precision Instrument Supplier No. EZ20027
The Euthanex Lid system has become an industry standard. Simply by removing the water bottle from the wire bar cage cover and placing the Euthanex Lid over the host cage, you are ready to perform euthanasia. You no longer need to transfer animals. The lids are available in five sizes to accommodate virtually all plastic cage sizes designed for small lab animals. These heavy duty stainless steel lids have a stem fitting for connection to the quick-disconnect fitting on the regulator hose from the regulator. A foam lid gasket ensures a good seal on the cage. Multiple lids may be used to treat several cages at once.