Learn More
Euthanex EUTHANEX LIDS SM SS 9X13
Catalog No. NC9903284
Encompass
Euthanex Lid, Small, 13 x 9 in. , Stainless steel, Fits old-style mouse cages, Each lid comes with one gasket and a plug hose stem
Catalog No. NC9903284 Supplier Euthanex Supplier No. E20027
Euthanex Lid, Small, 13 x 9 in. , Stainless steel, Fits old-style mouse cages, Each lid comes with one gasket and a plug hose stem