Learn More
Euthanex EP-1305 Low Flow CO2 Regulator for CO2 cylinders with CGA-320 fittings, 0.5 to 12 LPM Output.
Catalog No. NC9507464
Encompass
EP-1305 Low Flow CO2 Regulator, 0.5 to 12 LPM Output. Incorporates an adjustable flow meter and attaches directly to CO2 cylinder with CGA-320 fitting (optional CGA-940 Yoke connection available).Includes 9 foot hose with quick disconnect fitting.
Catalog No. NC9507464 Supplier Euthanex Supplier No. EP1305
EP-1305 Low Flow CO2 Regulator, 0.5 to 12 LPM Output. Incorporates an adjustable flow meter and attaches directly to CO2 cylinder with CGA-320 fitting (optional CGA-940 Yoke connection available).Includes 9 foot hose with quick disconnect fitting.