Learn More
System Biosciences LLC EV-GUARD STRG BUFFER 10XEV-GuardTM EV Storage Buffer (10X) = 4 mls
SDP
Catalog No. NC3552128
Encompass_Preferred
EV-GuardTM EV Storage Buffer (EV-GuardTM) is an innovative solution designed to safeguard Exosomes and maintain their biological activities during storage. xtracellular vesicles play a crucial role in cell-to-cell communication, carrying various biomolecules such as proteins, lipids, and nucleic acids. They have gained significant interest in research and clinical applications, such as diagnostics, therapeutics, and drug delivery. Preservation of the physical and biological characteristics of EVs is crucial for their use in downstream applications, making storage a key consideration.
Catalog No. NC3552128 Supplier System Biosciences LLC Supplier No. EXSBA10
EV-GuardTM EV Storage Buffer (EV-GuardTM) is an innovative solution designed to safeguard Exosomes and maintain their biological activities during storage. xtracellular vesicles play a crucial role in cell-to-cell communication, carrying various biomolecules such as proteins, lipids, and nucleic acids. They have gained significant interest in research and clinical applications, such as diagnostics, therapeutics, and drug delivery. Preservation of the physical and biological characteristics of EVs is crucial for their use in downstream applications, making storage a key consideration.