Learn More
Biotium EV stains Cell and Organelle Stains Exosome EV stains Chemical Reagents 5/PK
Catalog No. 5000501539
Encompass_Preferred
ExoBrite EV Stain Enhancer is an additive that can be added to extracellular vesicle staining reactions to improve signal to noise in flow cytometry by decreasing aggregates and increasing specific staining It is particularly beneficial for use with ExoBrite WGA and ExoBrite Annexin stains
Catalog No. 50-005-01539 Supplier Biotium Supplier No. 28002
ExoBrite EV Stain Enhancer is an additive that can be added to extracellular vesicle staining reactions to improve signal to noise in flow cytometry by decreasing aggregates and increasing specific staining It is particularly beneficial for use with ExoBrite WGA and ExoBrite Annexin stains