Learn More
Biotium EV stains Cell and Organelle Stains Exosome EV stains ExoBrite 400 460 1/EA
Catalog No. 5000501566
Encompass_Preferred
ExoBrite 400/460 True EV Membrane Stain is a membrane binding dye that stains EVs with full coverage and broad compatibility These dyes have improved solubility resulting in lower background and better signal to noise in EV detection compared to other membrane dyes 500X in DMSO 500 uL
Catalog No. 50-005-01566 Supplier Biotium Supplier No. 30136
ExoBrite 400/460 True EV Membrane Stain is a membrane binding dye that stains EVs with full coverage and broad compatibility These dyes have improved solubility resulting in lower background and better signal to noise in EV detection compared to other membrane dyes 500X in DMSO 500 uL