Learn More
Biotium EV stains Cell and Organelle Stains Exosome EV stains ExoBrite 410 450 1/EA
Catalog No. 5000501242
Encompass_Preferred
ExoBrite Annexin EV Staining Kits are designed for staining EVs and exosomes for detection by flow cytometry This optimized formulation of Annexin V binds to phosphatidylserine in the EV membrane with bright signal and low background Ex/Em 411/452 nm 500 labelings
Catalog No. 50-005-01242 Supplier Biotium Supplier No. 30119
ExoBrite Annexin EV Staining Kits are designed for staining EVs and exosomes for detection by flow cytometry This optimized formulation of Annexin V binds to phosphatidylserine in the EV membrane with bright signal and low background Ex/Em 411/452 nm 500 labelings